Tisk…
Ion selective electrode, ammonia
Dodavatel: Mettler - Toledo
51341000
662-2809EA 34930 CZK
662-2809
Ion selective electrode, ammonia
Elektrody Elektrody ISE
This gas-sensing electrode is intended for reliably measuring the ammonia content (NH₃).
  • Modular and versatile
  • Refillable electrolyte
  • Polymer matrix membrane