Tisk…
Pocket conductivity meters, LAQUAtwin, EC-11 / EC-22 / EC-33
Dodavatel: HORIBA
3999960125 3999960126 3999960127
662-2293EA 3900 CZK
662-2293 662-2294 662-2295
Pocket conductivity meters, LAQUAtwin, EC-11 / EC-22 / EC-33
Meřiče elektrochemické Konduktometry
These meters feature a unique flat sensor that enables quick, accurate and direct measurements of micro samples as small as 0,12 ml. The sensor does not need to be dipped into the sample, a few drops are sufficient.
  • Auto range change, automatic temperature compensation
  • Automatic hold, automatic calibration (2- or 3-point, depending on model)
  • Durable waterproof ABS housing, LCD display with backlight
  • Auto power off after 30 mins, low battery indicator
  • 400 hours battery life (continuous use without backlight)
  • Replaceable electrode
Available as threeMore Product Information