Tisk…
Ion selective electrodes, silver/sulfide, Orion™
Dodavatel: Thermo Orion
9416BN 9616BNWP
662-0834EA 22740 CZK
662-0834 662-1580
Ion selective electrodes, silver/sulfide, Orion™
Elektrody Elektrody ISE
These electrodes perform quick, simple, accurate and economical measurements of silver and sulfide ions in aqueous solutions. Ideal for performing low-level cyanide and halide titrations.
  • Sure-Flow™ reference junction prevents electrode clogging and provides fast and stabile readings
  • Available with a waterproof BNC ,BNC and screw cap connector and can be used on any ISE meter with a BNC connection
  • The free-flowing liquid junction assures stable, drift-free potentials
  • BNWP connector provides a waterproof and secure connection when used with the Orion Star