Tisk…

Rat IgG1 Isotype Control [clone: HRPN] (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®)

Dodavatel: Tonbo Biosciences
65-4301-U100
65-4301-U100EA 3580 CZK
65-4301-U100
Rat IgG1 Isotype Control [clone: HRPN] (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®)
Protilátky
The HRPN immunoglobulin is useful as an isotype-matched control. The HRPN immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG1 antibodies.

Type: Control
Antigen: IgG2b
Clonality: Monoclonal
Clone: HRPN
Conjugation: PerCP-Cy5.5®
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR