Tisk…
Brine tester
Dodavatel: Alla France
1120FG026/15QP
641-1500EA 797 CZK
641-1500
Brine tester
Hydrometry
Brine is the percentage of saturated solution of salt, usually sodium chloride, in water.
A brinometer is used to measure the percentage of saturated solution of brine.