Tisk…
Handheld refractometers, MASTER series
Dodavatel: Atago
2364 2374 2384 2363
635-0693EA 7900 CZK
635-0693 635-0697 635-0701 635-0692
Handheld refractometers, MASTER series
Refraktometry Refraktometry ruční
The MASTER series of handheld refractometers provides a range of easy to use instruments for a wide variety of applications.
  • MASTER-M: Standard model, clear scale, only two drops required
  • MASTER-T: Automated temperature compensation from 10 to +30 °C
  • MASTER-α: Automated temperature compensation (IP 65 except eyepiece), dust-tight and protected against water jets
  • MASTER-P: Designed with a plastic body for resistance to salty and acidic samples
The design has an ergonomic grip and a prism housing that allows for the sample to cover the prism withoutMore Product Information