Tisk…
Handheld refractometers, Eclipse
Dodavatel: BELLINGHAM STANLEY
45-41 45-44 45-26 45-01 45-02 45-03 45-05 45-06 45-08 45-22 45-46 45-65 45-81 45-82 45-66 45-006
635-0049EA 8210 CZK
635-0049 635-0050 635-0048 635-0037 635-0038 635-0039 635-0040 635-0041 635-0042 635-0044 635-0188 635-0291 635-0292 635-0293 BELL45-66 BELL45-006
Handheld refractometers, Eclipse
Refraktometry Refraktometry ruční
Handheld refractometers use high precision clear scale, and simple zero adjustment with lock.
  • Shaded field of view for distinction
  • Anti-roll support and ergonomic design
  • Comfortable eyepiece with smooth adjustment
  • Easy to clean prism and push-on flap protection
  • Unique sample ‘dribble’ feature
Informace o dodávce: Supplied with a soft carrying case and instructions.