Tisk…
Spectrophotometer cuvettes, NanoCuvette™ One
Dodavatel: cphnano
17 3
634-1176EA 0 CZK
634-1176 634-1177
Spectrophotometer cuvettes, NanoCuvette™ One
Kyvety Kyvety spektrofotometrické
Get accurate quantification by upgrading a UV-Vis spectrophotometer with user-friendly cuvettes. The NanoCuvette™ One expands the capabilities of an UV/Vis spectrophotometer to include quantification via refractive index for QC, kinetics, and 0,5 μl sample measurements.
  • Excellent for QC, kinetics, and label-free concentrations of a wide range of samples
  • All major UV/Vis spectrophotometer brands supported seamlessly out of the box
  • Based on semi-disposable PMMA cuvette, and reusable for low-cost measurements
  • Results independent of cleaning, pipetting errors,