Tisk…
VWR®, Kyvety jednorázové
Dodavatel: VWR Collection
634-0678EA 559 CZK
634-0678 634-0675 634-0676 634-0677
VWR®, Kyvety jednorázové
Kyvety Kyvety spektrofotometrické
Disposable cuvettes in either optical polystyrene (PS) or polymethyl methacrylate (PMMA), for use with most polar solvents, acids and alkaline solutions.
  • Minimum variation of extinction values
  • Recessed windows, clear and clean optical path
  • Optimum transmittance and reduced risk of contamination
For determinations in water analysis, chemistry and life science applications.
Balení: Resealable – cuvettes are assorted by cavity numbers.