Tisk…
Automatic transmittance colorimeters, PFX195 series, Lovibond®
Dodavatel: Lovibond Tintometer
1371951 1371952 1371953 1371954 1371955 1371957 1371958 1371959
633-0213EA 239590 CZK
633-0213 633-0214 633-0215 633-0216 633-0217 633-0219 633-0220 633-0221
Automatic transmittance colorimeters, PFX195 series, Lovibond®
Kolorimetry Automatické kolorimetry pro měření transmitance
A series of spectrophotometric colorimeters for colour grading of optically clear samples. Each version of the PFXi 195 includes a selection of standard colour scales that is used in a specific industry sector. Additional scales can be added, at the time of order or remotely, as optional upgrades, to provide the flexibility to meet individual requirements. Results can also be displayed in terms of spectral data and CIE values. For product types that are incompatible with the standard colour scales, the PFXi 195 software allows the users to build up a customised scale from a series ofMore Product Information