Tisk…
Comparator kits, Lovibond® system 2000
Dodavatel: Lovibond Tintometer
433280 433340 433320 433310 433330 433250 433300 433350
633-0022EA 56380 CZK
633-0022 633-0024 633-0023 633-0152 633-0153 633-0021 633-0151 633-0259
Comparator kits, Lovibond® system 2000
Kolorimetry Sady porovnávací
Available for many of the commonly used colour scales, these kits are a convenient means of ordering the complete range of equipment required. Each system includes.
  • A visual comparator instrument which brings the sample and colour standards into the same field of view for more accurate colour matching
  • A selection of colour standards - stable-coloured glass filters in test discs, which have been pre-calibrated to give direct readings over set measuring ranges
  • A standardised benchtop light source to ensure constant, uniform lighting conditions for colour
More Product Information