Tisk…
Slide staining system, StainTray™ M920
Dodavatel: Simport Scientific
M920-1 M921-1 M920-2 M921-2
631-1922EA 11400 CZK
631-1922 631-1924 631-1923 631-1925
Slide staining system, StainTray™ M920
Krabičky barvicí
StainTray M920 has a rigid base that is resistant to many chemicals (except chlorinated hydrocarbons). It can hold up to 20 slides on four plastic rails covered with a polymer strip for holding slides perfectly in place even if the tray is held at an angle.
  • Choice of two possible covers: Clear for visual examination/black for fluorescent work
  • Drain plug can be removed and emptied easily
  • Four rubber feet to increase base stability
  • Units are stackable to save space
  • Single-handed operation makes handling safe and easy
Suitable for routine stainingMore Product Information