Tisk…
VWR®, Box Top Parafilm® Dispenser
Dodavatel: VWR Collection
631-1280EA 1370 CZK
631-1280
VWR®, Box Top Parafilm® Dispenser
Zásobníky na fólie/parafilm
Easy application with no mess.
  • Accommodates two 50 mm (2"), or one 100 mm (4") Parafilm® M rolls
  • Made from ABS plastic, making the unit easy to clean
Easy application involves simply placing the dispenser on top of the Parafilm® M box and pulling the sealing film through. There is no need to remove rolls from the packaging. A built-in safety blade slides and cuts with no danger to fingertips, eliminating the need for scissors.
Informace pro objednávání: Supplied with two safety blades, Additional replacement blades are available separately for purchase.