Tisk…
VWR®, Silane Coated, Double Frosted Slides
Dodavatel: VWR Collection
631-1162EA 5740 CZK
631-1162
VWR®, Silane Coated, Double Frosted Slides
Sklíčka mikroskopická a krycí Sklíčka mikroskopická adhezivní
Silane (aminoalkylsilane) coating provide permanent adhesion, which can bind tissue sections to glass slides with a sticky surface.
  • Smooth edges for operational safety
  • Marking with pencils, marker pens and conventional labelling systems
  • Shelf life of 18 months
Recommended for H&E stain, immunohistochemistry (IHC) and hybridisation on frozen sections in histopathology examination.
Balení: 50 pieces per box. 50 boxes per case.
Upozornení: Fragile items should be stored in dry and proper laboratory temperature conditions.