Tisk…
Counting chambers, Imp. Neubauer
Dodavatel: LABOR OPTIK
1106000 1120000 1110000 1130000
631-1111EA 2580 CZK
631-1111 631-1121 631-1112 631-1113
Counting chambers, Imp. Neubauer
Sklíčka mikroskopická a krycí Komůrky počítací
For counting leucocytes, thrombocytes and erythrocytes. All group squares have triple border lines.
  • Four large squares in the corners are divided into 16 squares each (leucocyte counting)
  • Large square in the middle divided in 25 squares, each of which is further divided into 16 small squares
  • The middle line determines whether cells in the border area are to be included in one square or the other
Counting grid: 9
Large squares: 1 mm² each
Large squares: 1 mm² each