Tisk…
Microscope slides, Menzel Gläser, SuperFrost®
Dodavatel: Epredia
AG00008032E01 AG00008132E01 AG00008232E01 AG00008332E01 AG00008432E01
631-0705EA 260 CZK
631-0705 631-0706 631-0707 631-0708 631-0709
Microscope slides, Menzel Gläser, SuperFrost®
Sklíčka mikroskopická a krycí Sklíčka mikroskopická
These slides come with a light opaque coating in standard colours.
  • Ground 90° edges
  • Label areas for easy identification
  • Resistant to solvents, and to scratching or sticking when stacked together
Each colour codes different specimens at a glance according to urgency, processing or staining technique.