Tisk…
Upright microscopes, biological, Panthera C2
Dodavatel: MOTIC
1100104601132 1100104601142 1100104601131 1100104601141 1100104601162 1100104601172
630-3452EA 47980 CZK
630-3452 630-3454 630-3451 630-3453 630-3455 630-3456
Upright microscopes, biological, Panthera C2
Mikroskopy
Panthera C2, Motic's new upright microscope range, revolutionises the conventional microscope concept by creating a smart microscope. Its design, together with its new optics and digital capabilities, makes it the microscope of tomorrow. Your perfect lab mate.
  • Motic LightTracer: Light memory, sleep mode (auto on-off), nosepiece LED light intensity indicator unique rack less stage compact and with double-slide holder
  • Coded nosepiece - UC widefield eyepieces 10×/22 mm
  • Digital multifunctional knob
  • LED illumination, with a red LED strip on the revolver
More Product Information