Tisk…
Upright microscopes, DM 1000 LED
Dodavatel: LEICA MICROSYSTEMS
11888842_P1 11888842_P2 11888842_P3 11888842_P4 11888842_P5 11888842_P6 11888842_P7 11888842_P8
630-3168EA 158760 CZK
630-3168 630-3169 630-3170 630-3171 630-3172 630-3173 630-3174 630-3175
Upright microscopes, DM 1000 LED
Mikroskopy
The DM1000 LED laboratory microscope is perfect for clinical laboratory applications; histology, cytology, haematology and pathology.
  • Compatible with the full line of Leica microscope optics and accessories for increased flexibility
  • Choice of traditional Tungsten Halogen light source or LED illumination that makes changing lamps a thing of the past
  • Appropriately scaled footprint to meet the needs of even the most space challenged laboratory
  • Easily the best all round choice in microscopes for transmitted light applications; brightfield and phase contrast
More Product Information