Tisk…
Immediate comparison software, Trace
Dodavatel: MOTIC
1101001500722
630-3042EA 4320 CZK
630-3042
Immediate comparison software, Trace
Software Laboratory Software
Trace software enables you to connect two or more Moticams or several digital microscopes to your computer to give you live comparative imaging.
  • Transform your microscopes into comparison microscopes and economically enter the area of forensic science teaching
  • More than just image comparison, you can overlay images in real time and retouch them
  • Authenticate handwriting in a few seconds, or determine whether two pieces of fabric belong to the same batch
  • Easy to install and to use, Trace provides a complete platform for evaluating comparative imaging based
More Product Information