Tisk…
VWR®, IOS W-Plan Objectives
Dodavatel: VWR Collection
630-2949EA 1270 CZK
630-2949 630-2948 630-2950 630-2954 630-2955 630-2951 630-2952 630-2953
VWR®, IOS W-Plan Objectives
Objektivy mikroskopu
These high contrast objectives are ideal for routine observations in laboratories.
  • Ensure field flatness up to F.N. 22
  • Based on infinity-corrected optical system
  • 100× is also available with iris for darkfield