Tisk…
Stereo zoom microscopes, Stemi 305
Dodavatel: ZEISS
4350639010100 4350639020100 4350639030100 4919030006000
630-2624EA 40450 CZK
630-2624 630-2625 630-2626 ZEIS4919030006000
Stereo zoom microscopes, Stemi 305
Mikroskopy
Stemi 305 are compact Greenough stereo microscopes with 5:1 zoom for biological education, labs and industrial production environments.
  • Three-dimensional and crisp images
  • Integrated LED illuminations: Near vertical and oblique reflected light, as well as transmitted light
  • White LEDs generate bright light of day­light colour
  • Can easily be operated, also by untrained users
  • Trinocular model provides a phototube for connection to any ZEISS Axiocam microscope camera
Choice between predefined microscope sets for education, industrial production andMore Product Information