Tisk…
Microscope stand, precision, Dino-Lite, RK-10A
Dodavatel: Dino-Lite / IDCP B.V.
RK-10A RK-10-FX RK-10-EX RK-10-VX RK-10-PX2
630-2517EA 9570 CZK
630-2517 IDCPRK-10-FX IDCPRK-10-EX IDCPRK-10-VX IDCPRK-10-PX2
Microscope stand, precision, Dino-Lite, RK-10A
Stojany laboratorní
The Dino-Lite RK-10A is a sturdy and stable high-end stand that can be used with all professional Dino-Lite models. It is constructed of resilient stainless steel and lightweight aluminium and offers a very precise fine-focus adjustment as well as a quick release function.
  • Quick release button allows for fast and convenient vertical movement
  • Fine-focus adjustment knob is especially useful at high magnification
  • Adjustable safety stop to prevent the microscope from touching the specimen
  • Adjustable horizontal arm (RK-10-EX, included) has a 15 cm range with 360
More Product Information