Tisk…
Staining systems, EasyDip™
Dodavatel: Simport Scientific
M900-12AS M905-12DGY M900-12B M900-12G M900-12P M900-12W M900-12Y M906 M906-12AS
720-0791EA 4280 CZK
720-0791 720-0793 720-0792 720-0794 720-0795 720-0796 720-0797 630-1540 SIMPM906-12AS
Staining systems, EasyDip™
Krabičky barvicí
The slide staining system is made of acetal plastic.
  • Jars can be loosely joined to each other to keep the same order when moved around
  • Wide base for stability while inside is recessed to reduce reagent volume
  • Available in five colours to aid identification of contents or applications
The slide staining system has two components: A square staining jar and a 12 position vertical slide rack. The staining jar is made of resistant acetal plastic and will resist most staining agents including alcohol and xylene but not phenol. Autoclavable, it is thereforeMore Product Information