Tisk…
Digital thermometers, testo 905
Dodavatel: Testo
0560.9055 0560.9056
620-3313EA 2270 CZK
620-3313 620-3314
Digital thermometers, testo 905
Teploměry Teploměry digitální
Mini penetration and surface digital thermometers with rotatable display.
  • Automatic off function
  • Industrial sensor, K-type thermocouple
  • Easy readout of value thanks to the rotatable display
The testo 905 is a fast mini thermometer with a broad measuring range. The 905-T1 has a professional industrial sensor (thermocouple K-type), which has an excellent level of accuracy, especially in the upper measuring range. The sprung, wide thermocouple measuring head of the 905-T2 guarantees a very quick reaction time, and high accuracy level since it always liesMore Product Information