Tisk…
Air/infrared surface thermometer, testo 810
Dodavatel: Testo
0560.0810
620-3306EA 2600 CZK
620-3306
Air/infrared surface thermometer, testo 810
Teploměry Teploměry digitální
The testo 810 allows the measurement of air temperature and simultaneous non contact surface temperature measurement in one instrument. Come complete with illuminated display.
  • One point laser spot marking with 6:1 optics
  • Display of difference between air and surface temperature
  • 'HOLD' function
  • Maximum/minimum values
  • Adjustable emissivity
Integrated NTC air thermometer.
Informace o dodávce: Supplied with a factory calibration protocol, protective cap, wrist strap, belt holder and two AAA batteries for about 50 hours of use without display illumination.