Tisk…
Pocket thermo-hygrometer, testo 610
Dodavatel: Testo
0560.0610
620-3305EA 4330 CZK
620-3305
Pocket thermo-hygrometer, testo 610
Vlhkoměry Teploměry-vlhkoměry digitální
Very handy, small and easy to use, this thermo-hygrometer provides a long-term drift-free probe. It includes a backlit display and an automatic off function.
  • Dew point calculation
  • Wet bulb measurements
  • Long-term stable humidity sensor
  • 'HOLD' and maximum/minimum value functions
The handy testo 610 thermo-hygrometer proves to be an excellent measuring instrument when it comes to fast, precise measurement of temperature and humidity in indoor areas. In addition, the compact instrument also calculates the dew point and the wet bulb temperature as well.
Objednat
Produktová specifikace
Environmentally Preferable
Baterie 2 micro AAA type batteries 1,5 V (LR03)