Tisk…
VWR® Durac™ Plus, Hydrometers
Dodavatel: VWR Collection
620-2388EA 6050 CZK
620-2388 620-2389
VWR® Durac™ Plus, Hydrometers
Hydrometry
These plain form hydrometers are designed for applications where rough handling or contamination could be an issue.
  • Durable and shatterproof, lead free ballast
  • Easy to read black scale on yellow back to avoid eye fatigue
  • Individually serialised - Temperature standardisation of 15 °C
Suitable for applications in the food or beverage industries.
Certifikace: NIST.
Informace pro objednávání: Includes a multi-language statement of accuracy.
Upozornení: Not recommended for use with petroleum-based liquids or other corrosives, these should beMore Product Information