Tisk…
General-purpose thermometer
Dodavatel: SciLabware
MF31/2S
620-2290EA 5000 CZK
620-2290
General-purpose thermometer
Teploměry Analogue Thermometers
Spirit thermometers.
An integral ground glass cone on the stem allows direct fitting into Quickfit® jointed apparatus. Spirit thermometers have blue or green spirit for improved visibility.