Tisk…
VWR® Traceable®, Teploměr digitální
Dodavatel: VWR Collection
620-2023EA 2850 CZK
620-2023 620-1661
VWR® Traceable®, Teploměr digitální
Teploměry Teploměry digitální
These digital thermometers are suitable for monitoring of temperatures in freezers, water baths, heating blocks, incubators and refrigerators. The bottle-enclosed, temperature-buffered sensor is insulated from transient temperature changes. Micro cable allows refrigerator doors to close on it.
  • Maximum/minimum monitors high and low readings overnight, at the weekend or for any time period
  • Programmable alarm sounds continuously when temperature rises above/falls below set points and can be turned off manually
  • Visual and audible signals continue even if