Tisk…
VWR® RM 10, Moisture Tester
Dodavatel: VWR Collection
620-1956EA 4970 CZK
620-1956
VWR® RM 10, Moisture Tester
Analyzátory Analyzátory vlhkosti
The RM 10 allows detection of moisture behind ceramic tiles and/or various wall or floor coverings, in wood down to a depth of 3 cm and in concrete or wash floor down to 4 cm.
  • Easy and fast moisture rating, non destructive measurement
  • Displayed values by „digit“ are relative so that values can be compared
  • To check moisture on surfaces – No injection into the measuring object required
Suitable for high-speed determination of moisture i.e. estate agents, property management, house owners, architects, building experts, building contractors.
Upozornení:More Product Information