Tisk…
VWR®, Digital Thermo-Hygrometer
Dodavatel: VWR Collection
620-1951EA 5360 CZK
620-1951
VWR®, Digital Thermo-Hygrometer
Vlhkoměry Teploměry-vlhkoměry digitální
This handheld instrument is designed to measure temperature and humidity in air conditioning and ambient air monitoring. It provides a quick response in EDP rooms, galleries, offices, workshops, storage rooms, private buildings and greenhouses.
PT1000 and capacitive-polymer sensor (L×Ø: 35×14 mm).
Informace pro objednávání: Hygrometer is supplied with a test certificate and does not come with an ISO calibration certificate. If ISO calibration certificate is required, it has to specified during ordering. If calibration certificate is required after delivery, the unit has to be sent backMore Product Information