Tisk…
Thermo-hygrometers, 622 and 623
Dodavatel: Testo
0560.6230 0560.6220
620-1687EA 5470 CZK
620-1687 620-1686
Thermo-hygrometers, 622 and 623
Vlhkoměry Teploměry-vlhkoměry digitální
The model 622 measures the current temperature, humidity and air pressure. In addition, it displays date and time. The model 623 thermohygrometer measures the temperature and humidity in offices, living spaces, and commercial premises, and logs the readings for 90 days.
  • Table and wall mount available
  • A visual LED alarm alerts when room’s thresholds are reached
The testo model 622 digital hygrometer and barometer is ideal for monitoring the ambient air. The meter measures the air temperature, relative humidity and the air pressure at a rate of onceMore Product Information
Objednat
Produktová specifikace
Environmentally Preferable
Baterie 4×AA batteries, with 12 months of battery life
Materiál krytu ABS