Tisk…
Texture analysers, QTS 25
Dodavatel: AMETEK
QTS-25-05-230 QTS-25-25-230
619-0001EA 0 CZK
619-0001 619-0002
Texture analysers, QTS 25
Analyzátory Analyzátory Textury
The QTS-25 is a rugged benchtop instrument designed for the measurement of texture properties in production environments. It can operate in stand-alone mode using the included dedicated console or with a PC using the TexturePro software. In-process control testing applications in the food, cosmetic, pharmaceutical, polymers and textile industries.
  • Choice of 5 or 25 kg interchangeable load cells
  • 100 to 520 mm measuring distance accommodates most sizes
  • Range of probes and accessories for both compression and tension forces
Probes allow for theMore Product Information