Tisk…
Commercial grade hydrometers
Dodavatel: Alla France
303.100FRG 303.120FRG 370.087TRFCP 3011FC060/20QP 3011FC07020QP 301.080TRFRGP 301.090TRFRGP 301.100TRFRGP 301.110TRFRGP 301.120TRFRGP 301.130TRFRGP 301.140TRFRGP 301.150TRFRGP 301.160TRFRGP 301.170TRFRGP 301.180TRFRGP 3011FC190/20QP 302.065TRFRGP 302.080TRFRGP 302.100TRFRGP 302.120TRFRGP 3021FC140/20QP 302.160TRFRGP 370.093TRFCP 370.099TRFCP 370.105TRFCP 3700FC111/20QP 3700FC117/20QP 3700FC123/20QP 3700FC129/20QP 370.135TRFCP 3700FC141/20QP 370.147TRFCP
ALLA303.100FRGEA 1010 CZK
ALLA303.100FRG ALLA303.120FRG 614-1557 614-1503 614-1504 614-1505 614-1506 614-1507 614-1508 614-1509 614-1510 614-1511 614-1512 614-1513 614-1514 614-1515 614-1516 614-1517 614-1518 614-1519 614-1520 614-1521 614-1522 614-1558 614-1559 614-1560 614-1561 614-1562 614-1563 614-1564 614-1565 614-1566 614-1567
Commercial grade hydrometers
Hydrometry
Commercial grade hydrometers − Not standardised.
  • Reference mark on the stem
  • Check of the calibration on the paper scale
  • Bulb Ø is 15 or 20 mm with a stem Ø of 6 to 7 mm for greater accuracy