Tisk…
Axygen® LTS Compatible Pipette Tips
Dodavatel: Corning
LT-200-R LT-200-RL LT-200-R-S LT-200-TS LT-200-TS-S LT-20-R LT-20-RL LTF-20-R-S LTFER-1000-R-S LTFER-200-R-S LT-1000-R LT-1000-RL LT-1000-R-S LT-20-R-S LTF-200-R-S LTFER-20-R-S
613-7415EA 1690 CZK
613-7415 613-7417 613-7416 613-7418 613-7419 613-7412 613-7414 613-7421 613-7423 613-7425 613-7409 613-7411 613-7410 613-7413 613-7422 613-7424
Axygen® LTS Compatible Pipette Tips
Špičky pipetovací
The new Axygen® LTS compatible pipette tips are ergonomically designed to reduce discomfort and are optimised to work with Rainin LiteTouch™ pipettors. These tips maintain the quality users have come to expect from corning.
  • Graduation lines for quick accuracy checks
  • Ultra-clear USP class VI resin
  • Fine point delivery system ensures crisp sample dispensing
Flexible sealing area optimised to work with LTS pipettors enables less force to be used during pipetting.
Certifikace: ISO 13485, ISO 14001. RNase-free, DNase-free, human DNA-free, PCR inhibitor-free, and non-pyrogenic.