Tisk…
VWR® 12-Channel Reagent Reservoirs
Dodavatel: VWR Collection
613-7292EA 1980 CZK
613-7292 613-7298 613-7300 613-7309
VWR® 12-Channel Reagent Reservoirs
Zásobníky na reagencie
These 12-channel reservoirs are made from high-quality, chemical resistant PP. Designed for serial dilutions or pipetting individual reagents using up to 12-channel pipetters.
  • V-shaped basins facilitate easy withdrawal of sample solutions
  • Each channel holds up to 3 ml of solution and is individually numbered
  • Includes a tight-fitting lid
  • Individually wrapped packaging
  • Non-sterile or sterilised by irradiation, SAL 10⁻⁶ (ISO 11137)
  • Lot-certified, non-pyrogenic, DNase- and RNase-free
12-channel reagent reservoirs feature twelve 3 ml compartments, designedMore Product Information