Tisk…
VWR® Standard Line, Multi-Channel Pipettes, Mechanical, Variable Volume
Dodavatel: VWR Collection
613-7111EA 16640 CZK
613-7111 613-7112 613-7113 613-7114 613-7115 613-7116 613-7117 613-7118
VWR® Standard Line, Multi-Channel Pipettes, Mechanical, Variable Volume
Pipety Micropipettes
These lightweight Standard Line pipettes feature a revolutionary suspension system which allows the shaft to retract slightly when pressed against a row of pipette tips. Each shaft moves independently, resulting in an even sealing force being applied to all 8 or 12 tips. The suspension system not only ensures secure tip sealing, but also eliminates tips being jammed.
  • Easily adjustable ejector height system to accommodate virtually all brands of tips
  • Contoured shape of the handle, 360° manifold and soft spring system delivers smooth and effortless pipetting,
More Product Information