Tisk…
Eppendorf® Pipette Manager - Connected Pipette System
Dodavatel: EPPENDORF
1004000.001 4861000.970
613-6958EA 31130 CZK
613-6958 613-6959
Eppendorf® Pipette Manager - Connected Pipette System
Software Laboratory Software
Control Eppendorf's connected electronic pipettes via a dedicated WiFi for faster, more reproducible results. Remotely set up pipettes, receive pipetting technique guidance, digitally document activities, and more.
  • Connected electronic pipette system to remotely set up electronic pipettes, ease handling of challenging liquids and digitally document pipetting steps
  • Choose from predefined Eppendorf liquid types (e.g., ethanol or enzymes/antibodies) or create your own
  • Manage your pipette fleet more effectively and enhance collaboration with integrated