Tisk…
Single-channel pipettes, electronic, with Bluetooth, E1-ClipTip™
Dodavatel: Thermo Fisher Scientific
4670000BT 4670020BT 4670030BT 4670040BT
613-5748EA 0 CZK
613-5748 613-5749 613-5750 613-5751
Single-channel pipettes, electronic, with Bluetooth, E1-ClipTip™
Pipety Micropipettes
These pipettes are ideal for repetitive pipetting tasks with microcentrifuge or test tubes. Electronic tip ejection and index finger pipetting action allow the thumb to relax during pipetting.
  • Tip is locked and sealed in place until ejected − no dropped tips that waste valuable samples
  • Two operating options for every application: Step based Matrix or Pre-sets with settings for most common applications
  • Five user profiles, personalised user interface and password protection offer unified operation for multiple users and environments
  • Ability to store up to 30
More Product Information