Tisk…
Multi-packs, Proline® Plus
Dodavatel: Sartorius
LH-728670 LH-728671 LH-728672 LH-728673 LH-728674 LH-728675 LH-728676 LH-728677
613-5732EA 20070 CZK
613-5732 613-5733 613-5734 613-5735 613-5736 613-5737 613-5738 613-5739
Multi-packs, Proline® Plus
Pipety Micropipettes
These Multipacks combine a set of Proline® Plus mechanical pipettes with matching tip trays and useful accessories. They offer an opportunity to test and get started with Proline® Plus mechanical pipettes.
  • Ergonomic design with light pipetting forces, comfortable handle and finger support for reduced risk of strain injuries
  • Minimised risk of contamination with Safe-Cone Filters and full autoclavability
  • Highly durable with strengthened structure also for heavier use
It is the widest range of pipettes for various users and applications.
Informace oMore Product Information