Tisk…
Pipette tips, uTIP™, X-Resin™
Dodavatel: BIOTIX
M-0200-9SC M-1250-9SC 63300069 M-0200-9TN M-0300-9TN M-1250-9TN 63300077 M-0250-BC M-1000-9NC 63300081 63300048 63300049 63300050 63300052 63300054 63300056 63300059 63300055 63300061 63300057 63300064 63300070 63300071 63300073 63300075 63300076 63300078 63300080 63300082 63300051 63300058 63300072 63300079 63300053 63300060 63300074
613-1378EA 0 CZK
613-1378 613-1380 613-5616 613-5614 613-5613 613-5612 613-5623 613-5861 613-5863 613-5866 BITX63300048 613-1377 BITX63300050 613-1379 BITX63300054 613-2814 BITX63300059 613-2813 BITX63300061 613-2815 613-5619 613-5615 BITX63300071 BITX63300073 BITX63300075 613-5622 613-5624 BITX63300080 613-5647 BITX63300051 BITX63300058 BITX63300072 BITX63300079 613-5862 BITX63300060 613-5865
Pipette tips, uTIP™, X-Resin™
Špičky pipetovací
This new generation of tips form an excellent seal on both single- and multi-channel pipettes guaranteeing total recovery of the sample whilst protecting the technician against the risks of repetitive strain injury. The tips are manufactured with proprietary technologies for increased pipetting accuracy and precision.
  • Made from transparent PP
  • FlexFit™ provides a flexible nature at the proximal end of the tip to reduce the insertion and ejection forces when using a pipette
  • Blade™ technology eliminates hanging drops at the distal end of the tip
  • Certified
More Product Information
Objednat
Produktová specifikace
Environmentally Preferable Waste Reducing,Udržitelná balení