Tisk…
Pipette tips, FlexiBulk, Optifit
Dodavatel: Sartorius
LH-B790204 LH-B790354 LH-B791004 LH-B791024 LH-B791204
613-3887EA 1090 CZK
613-3887 613-3888 613-3889 613-3890 613-3891
Pipette tips, FlexiBulk, Optifit
Špičky pipetovací
The FlexiBulk concept combines the low cost of bulk tips with saving space, improving usability and securing tip purity.
  • Reduces up to 40% on pack volume; cuts packaging waste by nearly 50%
  • Saves time and effort: Well-organized FlexiBulk pack makes tip picking quicker and more convenient
  • Tips are certified as free of RNase, DNase and endotoxins
  • Tips are manufactured under ISO Class 8 cleanroom conditions
  • Lid provides an airtight seal during transport and storage for secured purity
  • Lid is easy to close between uses to protect the tips from dust