Tisk…
Automation tips for Tecan®, Axygen®
Dodavatel: Corning
TT-1000-CBKHTR TT-1000CBKHTRS TT-1000-C-HTR TT-1000-CHTRS TT-200-CBK-HTR TT-200-CBKHTRS TT-200-C-HTR TT-200-C-HTR-S TT-20-CBK-HTR TT-50-CBK-HTR TT-50-CBK-HTRS TT-50-C-HTR TTF1000CBKHTRS TTF-1000-CHTRS TTF-200CBKHTRS TTF-200-CHTRS TTF-20-CBKHTRS TTF-20-C-HTR-S TTF-50-CBKHTRS TTF-50-C-HTR-S TT1000CBKHTR24 EV-100-R EV-200-R EV-200-R-S EV-50-R TT50CBKHTR36 EV-50-R-S EVF-100-R-S EVF-180-R-S EVF-50-R-S EV-205-WB-R EVF-205-WB-R-S EV-200-NTR EV-50-NTR EV-200-NTR-S EV-50-NTR-S TTF900CBKHTRS
732-0961EA 5800 CZK
732-0961 732-0963 732-0960 732-0962 732-0970 732-0971 732-0968 732-0969 732-0966 732-0974 732-0975 732-0972 732-0983 732-0976 732-0980 732-0979 732-0978 732-0977 732-0982 732-0981 736-0485 736-0447 736-0449 736-0450 736-0451 736-0488 736-0452 736-0453 736-0454 736-0455 736-0523 736-0525 613-3882 613-3813 613-3812 613-3814 AXYGTTF900CBKHTRS
Automation tips for Tecan®, Axygen®
Špičky pipetovací
PP, with or without filter, sterile or non sterile.
  • Black tips are conductive for liquid level-sensing protocols
  • Optimised fine point for accurate dispensing
  • Certified RNase-, DNase- and endotoxin-free
These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
EVO-style tips are compatible with Tecan Freedom EVO®, Caliper Zephyr® SPE, and BioTek® Precision™ robotic pipetting systems.
Genesis-style tips are compatible with Tecan Genesis Freedom,More Product Information
Objednat
Produktová specifikace
Environmentally Preferable Low Manufacturing Impact
Certifikát ochrany životního prostředí I-RECs, ISO 14001 - Reynosa