Tisk…
Finntip™ Filtered Pipette Tips
Dodavatel: Thermo Fisher Scientific
94052000 94052020 94052100 94052150 94052200 94052320 94052350 94052450 94052310 94052430 94052060 94052550 94052160 94052410 94052300 94052800
613-2656EA 3869 CZK
613-2656 613-4490 613-2658 613-4492 613-2659 613-4494 613-2661 613-4497 613-4493 613-4496 613-4491 613-5011 613-5023 613-4495 613-2660 613-7087
Finntip™ Filtered Pipette Tips
Špičky pipetovací
Integral filter made from water-repellent PE which does not swell on accidental penetration by liquid, allowing recovery of the sample. Protect pipettes and the samples from contamination with Thermo Scientific™ Finntip™ Filtered Pipette Tips. These sterile, disposable pipette tips keep pipettes clean, and are perfect for PCR amplification applications and for pipetting radioactive, infectious, and aerosol-emitting samples. Available in racks with a wide volume range of 0,2 μl to 10 ml.
  • High quality PP with PE filter
  • Sterilised by gamma irradiation,
More Product Information