Tisk…
Pipette tips, low retention, ART™
Dodavatel: Thermo Fisher Scientific
3520-05 3550-05 3101-05-HR 3101-05-RT 3102-05-HR 3102-05-RT 3501-05-HR 3501-05-RT 3502-05-HR 3502-05-RT 3521-05-HR 3521-05-RT 3522-05-HR 3522-05-RT 3551-05-HR 3551-05-RT 3552-05-HR 3552-05-RT 3771-05-HR 3771-05-RT 3772-05-HR 3772-05-RT
732-0324EA 0 CZK
732-0324 732-0466 613-2446 613-2447 613-2450 613-2451 613-2458 613-2459 613-2462 613-2463 613-2470 613-2471 613-2474 613-2475 613-2489 613-2490 613-2493 613-2494 613-2497 613-2498 613-2776 613-2777
Pipette tips, low retention, ART™
Špičky pipetovací
Using advanced low-retention technology, the hydrophobic inner surface of these tips significantly reduces sample loss to adhesion and maximises sample recovery. The tips are suitable for use when working with the most difficult samples, such as radioactive compounds, viscous materials, fragile cell lines, viruses, difficult proteins and PCR reagents.
  • Non sterile tips are certified to be free of RNase, DNase, DNA, ATP, endotoxins, pyrogens and PCR inhibitors
  • Sterile tips are certified to be free of RNase, DNase, DNA, ATP, bioburden, endotoxins, pyrogens
More Product Information