Tisk…
Elektronické jednokanálové pipety Eppendorf® Xplorer® Plus
Dodavatel: EPPENDORF
4861000.708 4861000.716 4861000.724 4861000.732 4861000.740 4861000.759 4861000.720 4861000.736 4861000.710
613-2229EA 24170 CZK
613-2229 613-2230 613-2231 613-2232 613-2233 613-2234 613-6984 613-6985 613-6983
Elektronické jednokanálové pipety Eppendorf® Xplorer® Plus
Pipety Micropipettes
Eppendorf® Xplorer® plus electronic single-channel pipettes provide an accurate, precise, ergonomic liquid handling solution, featuring an intuitive interface, programmable modes, and advanced charging technology for reliable, efficient pipetting, enhancing reproducibility and minimising errors.
  • Multifunctional rocker: 'Up is up and down is down™' principle
  • Clearly arranged color display; no sub-menus; help function
  • Innovative rocker: When the tip is ejected, the piston automatically returns to zero position
  • ON/OFF switch: Switches the pipetter off during
More Product Information