Tisk…
Hypodermic needles, HENKE-JECT® for single use
Dodavatel: HENKE SASS WOLF
4710005016 4710007030 4710006030 4710004512 4710004020 4710016040 4710011040 4710009040 4710009025 4710006025 4710008040 4710008025 4710012040 4710007040 4710004012 4710004525 4710012050 4710003012 4710008050 4710011025 4710011030 4710015050 4710005025 4710005525 4710007025 4710007050 4710011050 4710008016 4710012025 4710016025 4710018050 4710018040 4710009050 8300021961 8300010721 8300018623 8300013707 8300021960
613-2016EA 230 CZK
613-2016 613-2019 613-2018 613-2014 613-2013 613-2032 613-2028 613-2025 613-2024 613-2017 613-2022 613-2021 613-2029 613-2020 613-2012 613-2015 613-2030 613-2011 613-2023 613-2026 613-2027 613-2031 613-2094 615-0040 613-3114 613-3117 613-3122 613-3118 613-3124 613-3127 613-3129 613-3128 613-3120 HSWA8300021961 HSWA8300010721 HSWA8300018623 HSWA8300013707 HSWA8300021960
Hypodermic needles, HENKE-JECT® for single use
Jehly ke stříkačkám
Sterile, disposable, hub and needle shield in PP, cannula in stainless steel. These needles are regularly mechanically analysed and experiences of clinical practice are fed back in order to continuously improve product quality and safety for enhanced patient comfort.
  • Precision lancet point according to ISO 7864 with exact, smooth bevel cut
  • Thin wall tubing, dry siliconisation
  • Five year sterility after date of sterilisation
Suitable for Luer and Luer lock.
Balení: Individually packed in blister packs with colour coding.