Tisk…
Single channel pipettes, mechanical, fixed / variable volume, Finnpipette™ F3
Dodavatel: Thermo Fisher Scientific
4640000 4640010 4640020 4640030 4640040 4640050 4640060 4640070 4640080 4640090 4650000 4650010 4650020 4650030 4650040 4650050 4650060 4650070 4650080 4650090 4650100 4640100
613-1459EA 0 CZK
613-1459 613-1460 613-1461 613-1462 613-1463 613-1464 613-1465 613-1466 613-1482 613-1483 613-1467 613-1468 613-1469 613-1470 613-1471 613-1472 613-1473 613-1474 613-1475 LABS4650090 LABS4650100 LABS4640100
Single channel pipettes, mechanical, fixed / variable volume, Finnpipette™ F3
Pipety Micropipettes
Superior comfort, performance, reliability and easiness of maintenance in one of the lightest pipettes available in the market.
  • Advanced Volume Gearing mechanism (AVG) - Self supportive modular mechanism increasing accuracy, precision and durability, with light force and smaller increments for volume adjustment
  • Large display with white numbers on black background for increased volume visibility and security
  • Double action pipetting button with rotating upper part preventing accidental volume alteration
  • Superior handle ergonomics and very low force pipetting
More Product Information