Tisk…
Finnpipette™ Novus GLP Kits
Dodavatel: Thermo Fisher Scientific
4701120 4701130 4700470 4700490
613-1428EA 0 CZK
613-1428 613-1429 613-5588 613-5589
Finnpipette™ Novus GLP Kits
Pipety Micropipettes
Access everything needed for pipetting in Finnpipette™ Novus GLP kits. These all-inclusive kits feature ergonomic and easy to use Finnpipette™ Novus pipetters. The kits contains four different volume range to choose from, and include Finnpipette™ Novus™ pipetters, appropriate stands and sample tips.
  • Greater ease-of-use and reduced risk of injury
  • Adjustable index-finger trigger action prevents common repetitive stress injuries caused by thumb-driven pipettes
  • Easy-to-follow prompts guide Novus users through 10 pipetting functions
All kits contain aMore Product Information