Tisk…
Pipette tips, Finntip™ Flex
Dodavatel: Thermo Fisher Scientific
94060510 94060513 94060517 94060520 94060810 94060813 94060820 94060710 94060713 94060717 94060720 94060100 94060103 94060117 94060120 94060817 94060970 94060973 94060310 94060313 94060317 94060320
613-0190EA 1808 CZK
613-0190 613-5019 613-5021 613-5022 613-0614 613-0615 613-0818 613-0811 613-0812 613-0814 613-0815 613-0806 613-0807 613-0809 613-0810 613-0817 613-0819 613-1598 613-0797 613-0798 613-0800 613-0801
Pipette tips, Finntip™ Flex
Špičky pipetovací
New generation of tips offering the user greater flexibility.
  • High quality PP, autoclavable
  • Ergonomic thin wall tip; low attachment and ejection forces
  • Non slip rack with hinged cover
  • Easy to use refill system
Balení: Available loose, in racks, in a starter kit (1 rack of 96 and 16 or 20 refills of 96) or a refill kit (16 or 20 refills of 96).
10 ml tips: only available in bags.